Learn More
Medchemexpress LLC D-ribose-13C | 70849-24-0 | MFCD00064325 | 99.9% | 151.12 g/mol | (13C)C4H10O5 | 100 MG
SDP
Catalog No. 5000300858
Encompass_Preferred
D-ribose-13C is a stable isotope-labeled form of D-ribose supplied as a white to off-white solid for laboratory research. It is used as a positional tracer in metabolic and biochemical studies to follow ribose incorporation and quantify metabolic flux by analytical techniques such as NMR and mass spectrometry. The material is high purity and packaged in small quantities for research-scale experiments.
  • 13C-labeled at carbon-1 for positional tracing.
  • High purity suitable for analytical applications.
  • White to off-white solid form for easy handling and storage.
  • Compatible with NMR and mass spectrometry workflows.
  • Available in small package sizes for research-scale experiments.
  • Intended for research use only; not for human consumption.
Catalog No. 50-003-00858 Supplier Medchemexpress LLC Supplier No. HYW018772S1100MG
D-ribose-13C is a stable isotope-labeled form of D-ribose supplied as a white to off-white solid for laboratory research. It is used as a positional tracer in metabolic and biochemical studies to follow ribose incorporation and quantify metabolic flux by analytical techniques such as NMR and mass spectrometry. The material is high purity and packaged in small quantities for research-scale experiments.
  • 13C-labeled at carbon-1 for positional tracing.
  • High purity suitable for analytical applications.
  • White to off-white solid form for easy handling and storage.
  • Compatible with NMR and mass spectrometry workflows.
  • Available in small package sizes for research-scale experiments.
  • Intended for research use only; not for human consumption.