Learn More
Medchemexpress LLC D-ribose-13C5 | 202114-47-4 | 100.0% | 155.09 g/mol | 13C5H10O5 | 100 MG
SDP
Catalog No. 5000301890
Encompass_Preferred
D-Ribose (mixture of isomers) -13C5 is a uniformly 13C-labeled D-ribose supplied as a white to off-white solid for use in metabolic tracing, stable-isotope labeling, and mass spectrometry applications. Packaged at 100 mg with high chemical and isotopic purity, it is intended for metabolic flux analysis and biochemical pathway studies.
  • Uniform 13C labeling for accurate metabolic tracing
  • High chemical purity and isotopic enrichment
  • White to off-white solid, easy to handle and weigh
  • Stable under recommended storage conditions for long-term use
  • Packaged quantity suitable for analytical and preparative workflows
Catalog No. 50-003-01890 Supplier Medchemexpress LLC Supplier No. HYW018772S15100MG
D-Ribose (mixture of isomers) -13C5 is a uniformly 13C-labeled D-ribose supplied as a white to off-white solid for use in metabolic tracing, stable-isotope labeling, and mass spectrometry applications. Packaged at 100 mg with high chemical and isotopic purity, it is intended for metabolic flux analysis and biochemical pathway studies.
  • Uniform 13C labeling for accurate metabolic tracing
  • High chemical purity and isotopic enrichment
  • White to off-white solid, easy to handle and weigh
  • Stable under recommended storage conditions for long-term use
  • Packaged quantity suitable for analytical and preparative workflows