Learn More
Medchemexpress LLC D-ribose-13C5 | 202114-47-4 | MFCD28346972 | 99.9% | 155.09 g·mol⁻1 | 13C5H10O5 | 1 MG
SDP
Catalog No. 5000300036
Encompass_Preferred
D-Ribose (13C5) is a stable-isotope labeled form of D-ribose supplied for analytical, tracer, and metabolic studies. It is provided as a high-purity solid suitable for use as an internal standard or metabolic labeling reagent, with documented solubility in water and recommended cold storage for long-term stability.
  • Stable-isotope labeled D-ribose for analytical and tracer applications.
  • High isotopic and chemical purity, typically ≥99.9%.
  • Soluble in water; sonication may be required for complete dissolution.
  • Available in small, research-scale quantities for method development.
  • Store as powder at -20°C for long-term stability.
Catalog No. 50-003-00036 Supplier Medchemexpress LLC Supplier No. HYW018772S151MG
D-Ribose (13C5) is a stable-isotope labeled form of D-ribose supplied for analytical, tracer, and metabolic studies. It is provided as a high-purity solid suitable for use as an internal standard or metabolic labeling reagent, with documented solubility in water and recommended cold storage for long-term stability.
  • Stable-isotope labeled D-ribose for analytical and tracer applications.
  • High isotopic and chemical purity, typically ≥99.9%.
  • Soluble in water; sonication may be required for complete dissolution.
  • Available in small, research-scale quantities for method development.
  • Store as powder at -20°C for long-term stability.