Learn More
Medchemexpress LLC D-Ribose, 5-(dihydrogen phosphate), sodium salt (1:2) | 18265-46-8 | 99.96% | 274.07 | 100 MG
SDP
Catalog No. 5000366600
Encompass_Preferred
D-Ribose 5-phosphate disodium is an intermediate of the oxidative branch of the pentose phosphate pathway (PPP) and an end product of the nonoxidative branch of the PPP. It is used in the synthesis of nucleotides and nucleic acids.
  • Human endogenous metabolite
Catalog No. 50-003-66600 Supplier Medchemexpress LLC Supplier No. HYW009371100MG
D-Ribose 5-phosphate disodium is an intermediate of the oxidative branch of the pentose phosphate pathway (PPP) and an end product of the nonoxidative branch of the PPP. It is used in the synthesis of nucleotides and nucleic acids.
  • Human endogenous metabolite