Learn More
Medchemexpress LLC D-Ribose 5-phosphate disodium dihydrate | 207671-46-3 | 99.0% | 310.10 | 1 ML
SDP
Catalog No. 5000366291
Encompass_Preferred
D-Ribose 5-phosphate disodium dihydrate is an intermediate of the oxidative branch of the pentose phosphate pathway (PPP) and an end product of the nonoxidative branch of the PPP.
  • Used in the synthesis of nucleotides and nucleic acids.
  • Generally assumed to arise via the oxidative and nonoxidative pentose phosphate pathways.
Catalog No. 50-003-66291 Supplier Medchemexpress LLC Supplier No. HYW009371C10MM/1ML
D-Ribose 5-phosphate disodium dihydrate is an intermediate of the oxidative branch of the pentose phosphate pathway (PPP) and an end product of the nonoxidative branch of the PPP.
  • Used in the synthesis of nucleotides and nucleic acids.
  • Generally assumed to arise via the oxidative and nonoxidative pentose phosphate pathways.