Learn More
Medchemexpress LLC D-Ribose 5-phosphate disodium dihydrate | 207671-46-3 | >99.0% | 310.10 | 100 MG
SDP
Catalog No. 5000453510
Encompass_Preferred
D-Ribose 5-phosphate disodium dihydrate is an intermediate of the oxidative branch of the pentose phosphate pathway (PPP) and an end product of the nonoxidative branch of the PPP. It is used in the synthesis of nucleotides and nucleic acids. Ribose-5-phosphate is generally assumed to arise via the oxidative and nonoxidative pentose phosphate pathways.
  • Endogenous metabolite
  • For research use only
  • Not sold to patients
Catalog No. 50-004-53510 Supplier Medchemexpress LLC Supplier No. HYW009371C100MG
D-Ribose 5-phosphate disodium dihydrate is an intermediate of the oxidative branch of the pentose phosphate pathway (PPP) and an end product of the nonoxidative branch of the PPP. It is used in the synthesis of nucleotides and nucleic acids. Ribose-5-phosphate is generally assumed to arise via the oxidative and nonoxidative pentose phosphate pathways.
  • Endogenous metabolite
  • For research use only
  • Not sold to patients