Learn More
Apexbio Technology LLC D-Ribose 50-69-1 100mg
SDP
Catalog No. 5000090789
Encompass_Preferred
D-Ribose (CAS 50-69-1) is a naturally occurring pentose monosaccharide involved in the pentose phosphate pathway where it serves as a crucial substrate for the biosynthesis of nucleotides and nucleic acids By facilitating the regeneration of adenosine triphosphate (ATP) D-Ribose supports cellular energy metabolism particularly under conditions of increased energy demand or compromised ATP production Preclinical and clinical research including ongoing trials examining the effects of D-Ribose on exercise performance investigate its capacity to enhance cellular bioenergetics D-Ribose is widely utilized in studies related to metabolic function myocardial energy balance and recovery from exercise-induced fatigue
Catalog No. 50-000-90789 Supplier Apexbio Technology LLC Supplier No. C7129100
D-Ribose (CAS 50-69-1) is a naturally occurring pentose monosaccharide involved in the pentose phosphate pathway where it serves as a crucial substrate for the biosynthesis of nucleotides and nucleic acids By facilitating the regeneration of adenosine triphosphate (ATP) D-Ribose supports cellular energy metabolism particularly under conditions of increased energy demand or compromised ATP production Preclinical and clinical research including ongoing trials examining the effects of D-Ribose on exercise performance investigate its capacity to enhance cellular bioenergetics D-Ribose is widely utilized in studies related to metabolic function myocardial energy balance and recovery from exercise-induced fatigue