Learn More
Medchemexpress LLC D-ribose-d2 | 478506-32-0 | 98.0% | 152.14 g·mol⁻1 | C5H8D2O5 | 5 MG
SDP
Catalog No. 5000300709
Encompass_Preferred
Deuterium-labeled D-ribose supplied as a white to off-white solid for use as an isotope-labeled standard and tracer in metabolic, biochemical, and analytical studies. Provided in a small, analytical package and stable when stored frozen for long-term use.
  • Deuterium-labeled tracer for metabolic flux analysis and tracer studies.
  • High chemical purity (98.0%) for reliable analytical results.
  • Suitable for mass spectrometry and NMR standards.
  • White to off-white solid, convenient small-scale packaging for analytical workflows.
  • Store frozen to maintain stability and shelf life.
Catalog No. 50-003-00709 Supplier Medchemexpress LLC Supplier No. HYW018772S105MG
Deuterium-labeled D-ribose supplied as a white to off-white solid for use as an isotope-labeled standard and tracer in metabolic, biochemical, and analytical studies. Provided in a small, analytical package and stable when stored frozen for long-term use.
  • Deuterium-labeled tracer for metabolic flux analysis and tracer studies.
  • High chemical purity (98.0%) for reliable analytical results.
  • Suitable for mass spectrometry and NMR standards.
  • White to off-white solid, convenient small-scale packaging for analytical workflows.
  • Store frozen to maintain stability and shelf life.