Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-ribose-d2 | 478506-32-0 | MFCD08459971 | 98.0% | 152.14 g/mol | C5H8D2O5 | 100 MG
SDP

Catalog No. 5000300830
Encompass_Preferred

D-ribose-d2 is a deuterium-labeled analogue of D-ribose (C5H8D2O5) provided for laboratory research. It supplies two deuterium atoms in the ribose moiety and is intended for tracer experiments, metabolic labeling, and analytical chemistry applications where stable-isotope incorporation aids detection and quantification.

  • Deuterium-labeled ribose for tracer and metabolic studies.
  • Facilitates detection and quantification by mass spectrometry.
  • Molecular formula C5H8D2O5; molecular weight 152.14 g/mol.
  • High purity (98.0%) suitable for analytical use.
  • Supplied in research-scale packaging (100 mg vial).

Catalog No. 50-003-00830 Supplier Medchemexpress LLC Supplier No. HYW018772S10100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-ribose-d2 is a deuterium-labeled analogue of D-ribose (C5H8D2O5) provided for laboratory research. It supplies two deuterium atoms in the ribose moiety and is intended for tracer experiments, metabolic labeling, and analytical chemistry applications where stable-isotope incorporation aids detection and quantification.

  • Deuterium-labeled ribose for tracer and metabolic studies.
  • Facilitates detection and quantification by mass spectrometry.
  • Molecular formula C5H8D2O5; molecular weight 152.14 g/mol.
  • High purity (98.0%) suitable for analytical use.
  • Supplied in research-scale packaging (100 mg vial).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.