Learn More
Medchemexpress LLC D-ribose-d2 | 478506-32-0 | MFCD08459971 | 98.0% | 152.14 g/mol | C5H8D2O5 | 50 MG
SDP
Catalog No. 5000301087
Encompass_Preferred
D-Ribose-d2 is a deuterium-labeled form of D-ribose (two deuterium atoms) used as a stable isotope reagent in metabolic tracing, NMR, and analytical research. It is supplied as a white to off-white solid with high purity and is intended for use in tracer studies and structural analysis. CAS 478506-32-0; molecular weight 152.14 g/mol.
  • Deuterium-labeled tracer for metabolic and flux studies.
  • Suitable for NMR and mass spectrometry experiments.
  • High purity for reliable analytical results.
  • White to off-white solid form for easy handling.
  • Stable when stored under recommended conditions.
  • Offered in small research-scale quantities.
Catalog No. 50-003-01087 Supplier Medchemexpress LLC Supplier No. HYW018772S1050MG
D-Ribose-d2 is a deuterium-labeled form of D-ribose (two deuterium atoms) used as a stable isotope reagent in metabolic tracing, NMR, and analytical research. It is supplied as a white to off-white solid with high purity and is intended for use in tracer studies and structural analysis. CAS 478506-32-0; molecular weight 152.14 g/mol.
  • Deuterium-labeled tracer for metabolic and flux studies.
  • Suitable for NMR and mass spectrometry experiments.
  • High purity for reliable analytical results.
  • White to off-white solid form for easy handling.
  • Stable when stored under recommended conditions.
  • Offered in small research-scale quantities.