Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-ribose-d5 | 99.1% | 155.16 g·mol⁻1 | C5H5D5O5 | 1 MG
SDP

Catalog No. 5000301364
Encompass_Preferred

D-Ribose-d5 is the deuterium-labeled form of D-ribose intended for use as a stable isotope reagent in metabolic, tracer, and analytical applications.

  • Deuterium-labeled (d5) ribose for tracer and metabolic studies.
  • High purity (99.10%) suitable for analytical applications.
  • Molecular formula C5H5D5O5; molecular weight 155.16 g·mol⁻1.
  • Useful as an internal standard for mass spectrometry and NMR.
  • Supplied in small mg-scale quantities for research use.

Catalog No. 50-003-01364 Supplier Medchemexpress LLC Supplier No. HYW018772S141MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Ribose-d5 is the deuterium-labeled form of D-ribose intended for use as a stable isotope reagent in metabolic, tracer, and analytical applications.

  • Deuterium-labeled (d5) ribose for tracer and metabolic studies.
  • High purity (99.10%) suitable for analytical applications.
  • Molecular formula C5H5D5O5; molecular weight 155.16 g·mol⁻1.
  • Useful as an internal standard for mass spectrometry and NMR.
  • Supplied in small mg-scale quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.