Learn More
Medchemexpress LLC D-ribose-d5 | 99.1% | 155.16 g/mol | C5H5D5O5 | 10 MG
SDP
Catalog No. 5000300035
Encompass_Preferred
D-ribose-d5 is the deuterium-labeled form of D-ribose used as a stable isotope tracer and internal standard for metabolic and analytical studies. It is suited for quantitative NMR, GC-MS, and LC-MS workflows and supports metabolic profiling and pharmacokinetic investigations.
  • Deuterium-labeled (d5) analog for isotope tracing.
  • Useful as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Facilitates metabolic profiling and pharmacokinetic studies.
  • High listed purity of 99.1%.
  • Available in small packaging sizes from 1 mg to 100 mg.
  • Powder stable at -20°C for long-term storage.
Catalog No. 50-003-00035 Supplier Medchemexpress LLC Supplier No. HYW018772S1410MG
D-ribose-d5 is the deuterium-labeled form of D-ribose used as a stable isotope tracer and internal standard for metabolic and analytical studies. It is suited for quantitative NMR, GC-MS, and LC-MS workflows and supports metabolic profiling and pharmacokinetic investigations.
  • Deuterium-labeled (d5) analog for isotope tracing.
  • Useful as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Facilitates metabolic profiling and pharmacokinetic studies.
  • High listed purity of 99.1%.
  • Available in small packaging sizes from 1 mg to 100 mg.
  • Powder stable at -20°C for long-term storage.