Learn More
Medchemexpress LLC D-ribose-d6 | 99.9% | 156.17 g/mol | C5H4D6O5 | 10 MG
SDP
Catalog No. 5000302032
Encompass_Preferred
D-Ribose-d6 is the deuterium-labeled analogue of D-ribose used as an isotope-labeled standard and tracer in biochemical and analytical studies. It facilitates mass spectrometry and NMR analysis of carbohydrate metabolism and energy-related pathways.
  • Deuterium-labeled tracer for mass spectrometry and NMR.
  • High isotopic enrichment suitable for quantitative analyses.
  • High chemical purity for analytical applications.
  • Supplied as a dry powder for easy handling and storage.
  • Useful as an internal standard or tracer in metabolic studies.
Catalog No. 50-003-02032 Supplier Medchemexpress LLC Supplier No. HYW018772S1210MG
D-Ribose-d6 is the deuterium-labeled analogue of D-ribose used as an isotope-labeled standard and tracer in biochemical and analytical studies. It facilitates mass spectrometry and NMR analysis of carbohydrate metabolism and energy-related pathways.
  • Deuterium-labeled tracer for mass spectrometry and NMR.
  • High isotopic enrichment suitable for quantitative analyses.
  • High chemical purity for analytical applications.
  • Supplied as a dry powder for easy handling and storage.
  • Useful as an internal standard or tracer in metabolic studies.