Learn More
Medchemexpress LLC D-ribose (mixture of isomers)-13C5 | 202114-47-4 | 99.9% | 155.09 g/mol | 13C5H10O5 | 5 MG
SDP
Catalog No. 5000300859
Encompass_Preferred
D-Ribose (mixture of isomers)-13C5 is the fully 13C-labeled form of D-ribose supplied as an isotopic standard for metabolic tracing and analytical workflows. It is intended as a reference material for mass spectrometry, quantitative assays, and experiments that require an isotopically labeled sugar standard.
  • Fully 13C-labeled D-ribose for isotopic studies.
  • High chemical purity (99.9%).
  • Soluble in water (100 mg/mL); ultrasonic assistance may be required.
  • Stable as a powder at -20°C (3 years) or 4°C (2 years).
  • Suitable as an analytical reference for mass spectrometry and metabolic tracing.
Catalog No. 50-003-00859 Supplier Medchemexpress LLC Supplier No. HYW018772S155MG
D-Ribose (mixture of isomers)-13C5 is the fully 13C-labeled form of D-ribose supplied as an isotopic standard for metabolic tracing and analytical workflows. It is intended as a reference material for mass spectrometry, quantitative assays, and experiments that require an isotopically labeled sugar standard.
  • Fully 13C-labeled D-ribose for isotopic studies.
  • High chemical purity (99.9%).
  • Soluble in water (100 mg/mL); ultrasonic assistance may be required.
  • Stable as a powder at -20°C (3 years) or 4°C (2 years).
  • Suitable as an analytical reference for mass spectrometry and metabolic tracing.