Learn More
Medchemexpress LLC D-ribose (mixture of isomers) -13C5 | 202114-47-4 | 99.9% | 155.09 g/mol | 13C5H10O5 | 50 MG
SDP
Catalog No. 5000300710
Encompass_Preferred
Uniformly 13C-labeled D-ribose (mixture of isomers) is supplied as a white to off-white solid for use as an isotope-labeled internal standard or tracer in metabolic profiling and analytical and mass-spectrometry applications. The compound (formula 13C5H10O5, CAS 202114-47-4) is provided in small research quantities with high isotopic enrichment and documented storage recommendations.
  • Uniform 13C labeling (13C5) for accurate tracing and quantitation.
  • High isotopic enrichment and purity (~99.9%).
  • White to off-white solid suitable for analytical workflows.
  • Available in small, measured sizes for tracer and standard use.
  • Stable when stored as powder at -20°C (3 years) or 4°C (2 years).
  • Compatible as an internal standard for mass spectrometry and metabolomics.
Catalog No. 50-003-00710 Supplier Medchemexpress LLC Supplier No. HYW018772S1550MG
Uniformly 13C-labeled D-ribose (mixture of isomers) is supplied as a white to off-white solid for use as an isotope-labeled internal standard or tracer in metabolic profiling and analytical and mass-spectrometry applications. The compound (formula 13C5H10O5, CAS 202114-47-4) is provided in small research quantities with high isotopic enrichment and documented storage recommendations.
  • Uniform 13C labeling (13C5) for accurate tracing and quantitation.
  • High isotopic enrichment and purity (~99.9%).
  • White to off-white solid suitable for analytical workflows.
  • Available in small, measured sizes for tracer and standard use.
  • Stable when stored as powder at -20°C (3 years) or 4°C (2 years).
  • Compatible as an internal standard for mass spectrometry and metabolomics.