Learn More
Medchemexpress LLC D-ribose (mixture of isomers) -13C5 | 202114-47-4 | MFCD28346972 | 99.9% | 155.09 g/mol | 13C5H10O5 | 10 MG
SDP
Catalog No. 5000301892
Encompass_Preferred
D-ribose (mixture of isomers) -13C5 is a fully 13C-labeled form of D-ribose intended for use as a stable isotope tracer in metabolic and analytical research. Supplied in milligram quantities and characterized for use in labeling studies and quantitative assays, it supports accurate detection and tracing in biochemical workflows.
  • Fully 13C-labeled on all five carbons for metabolic tracing.
  • High reported purity suitable for analytical applications.
  • Supplied in small, mg-scale quantities for research use.
  • Water soluble and compatible with common analytical workflows.
  • Useful for metabolic flux analysis and isotope labeling experiments.
Catalog No. 50-003-01892 Supplier Medchemexpress LLC Supplier No. HYW018772S1510MG
D-ribose (mixture of isomers) -13C5 is a fully 13C-labeled form of D-ribose intended for use as a stable isotope tracer in metabolic and analytical research. Supplied in milligram quantities and characterized for use in labeling studies and quantitative assays, it supports accurate detection and tracing in biochemical workflows.
  • Fully 13C-labeled on all five carbons for metabolic tracing.
  • High reported purity suitable for analytical applications.
  • Supplied in small, mg-scale quantities for research use.
  • Water soluble and compatible with common analytical workflows.
  • Useful for metabolic flux analysis and isotope labeling experiments.