Learn More
Medchemexpress LLC D-ribose (mixture of isomers), 13C5-labeled | 202114-47-4 | >98.0% | 155.13 g/mol | C5(13C5)H10O5 | 25 MG
SDP
Catalog No. 5000303958
Encompass_Preferred
D-ribose (mixture of isomers)-13C5 is a fully 13C-labeled stable isotope of D-ribose supplied as a mixture of isomers for use as an internal standard or tracer in metabolic and analytical studies. It is provided at high chemical purity and available in small milligram-scale packages suitable for analytical workflows.
  • Fully 13C-labeled on all five carbon atoms (13C5)
  • Supplied as a mixture of isomers
  • Chemical purity >98.0%
  • Molecular weight 155.13 g/mol
  • Suitable for use as an internal standard and tracer in metabolic and analytical studies
Catalog No. 50-003-03958 Supplier Medchemexpress LLC Supplier No. 25MGHYW018772S15
D-ribose (mixture of isomers)-13C5 is a fully 13C-labeled stable isotope of D-ribose supplied as a mixture of isomers for use as an internal standard or tracer in metabolic and analytical studies. It is provided at high chemical purity and available in small milligram-scale packages suitable for analytical workflows.
  • Fully 13C-labeled on all five carbon atoms (13C5)
  • Supplied as a mixture of isomers
  • Chemical purity >98.0%
  • Molecular weight 155.13 g/mol
  • Suitable for use as an internal standard and tracer in metabolic and analytical studies