Learn More
Medchemexpress LLC D-ribose (mixture of isomers) | 50-69-1 | MFCD00135453 | 98.0% | 150.13 | C5H10O5 | 250 MG
SDP
Catalog No. 5000302820
Encompass_Preferred
D-ribose (mixture of isomers) is a biologically relevant pentose sugar supplied as a white to off-white solid for laboratory research. It has a molecular formula of C5H10O5 and a molecular weight of 150.13 g/mol, with high solubility in water and DMSO and defined storage recommendations for stability.
  • High purity (98.0%) suitable for research applications.
  • White to off-white solid form, convenient for weighing and formulation.
  • Highly soluble in water (100 mg/mL) and DMSO (≥100 mg/mL) for easy preparation of stock solutions.
  • Stable under recommended storage: powder -20°C for long-term preservation.
  • Suitable for metabolic studies, enzymatic assays, and cell culture supplementation.
Catalog No. 50-003-02820 Supplier Medchemexpress LLC Supplier No. 250MGHYW018772
D-ribose (mixture of isomers) is a biologically relevant pentose sugar supplied as a white to off-white solid for laboratory research. It has a molecular formula of C5H10O5 and a molecular weight of 150.13 g/mol, with high solubility in water and DMSO and defined storage recommendations for stability.
  • High purity (98.0%) suitable for research applications.
  • White to off-white solid form, convenient for weighing and formulation.
  • Highly soluble in water (100 mg/mL) and DMSO (≥100 mg/mL) for easy preparation of stock solutions.
  • Stable under recommended storage: powder -20°C for long-term preservation.
  • Suitable for metabolic studies, enzymatic assays, and cell culture supplementation.