Learn More
Medchemexpress LLC D-ribose (mixture of isomers) | 50-69-1 | MFCD00135453 | 98.0% | 150.13 g/mol | C5H10O5 | 1 G
SDP
Catalog No. 5000301086
Encompass_Preferred
D-Ribose (mixture of isomers) is a naturally occurring pentose sugar supplied as a white crystalline powder for research use. It functions as an energy metabolite and enzyme substrate and is commonly used in studies of nucleotide synthesis, metabolic pathways, and protein glycation.
  • High purity (98.0% by NMR).
  • Molecular weight 150.13 g/mol; chemical formula C5H10O5.
  • Suitable as an enzyme substrate and metabolite in biochemical assays.
  • White crystalline powder with good water solubility.
  • Provided in laboratory-scale pack sizes for research applications.
Catalog No. 50-003-01086 Supplier Medchemexpress LLC Supplier No. HYW0187721G
D-Ribose (mixture of isomers) is a naturally occurring pentose sugar supplied as a white crystalline powder for research use. It functions as an energy metabolite and enzyme substrate and is commonly used in studies of nucleotide synthesis, metabolic pathways, and protein glycation.
  • High purity (98.0% by NMR).
  • Molecular weight 150.13 g/mol; chemical formula C5H10O5.
  • Suitable as an enzyme substrate and metabolite in biochemical assays.
  • White crystalline powder with good water solubility.
  • Provided in laboratory-scale pack sizes for research applications.