Learn More
Medchemexpress LLC D-ribose (mixture of isomers) | 50-69-1 | MFCD00135453 | 98.0% | 150.13 g/mol | C5H10O5 | 1 ML
SDP
Catalog No. 5000301085
Encompass_Preferred
D-ribose (mixture of isomers) is a naturally occurring five-carbon sugar and endogenous metabolite that serves as the sugar moiety of ATP. Supplied for research use, it supports metabolic and biochemical studies and is provided in both ready-to-use solution and solid formats to simplify assay preparation and analytical workflows.
  • High purity (98.0%) suitable for research applications.
  • Available as 10 mM solution in DMSO (1 mL) for convenient use.
  • Also available in solid quantities (100 mg-1 g) and bulk sizes on request.
  • Molecular weight 150.13 g/mol; formula C5H10O5.
  • Useful in studies of ATP metabolism, energy pathways, and protein glycation.
Catalog No. 50-003-01085 Supplier Medchemexpress LLC Supplier No. HYW01877210MM/1ML
D-ribose (mixture of isomers) is a naturally occurring five-carbon sugar and endogenous metabolite that serves as the sugar moiety of ATP. Supplied for research use, it supports metabolic and biochemical studies and is provided in both ready-to-use solution and solid formats to simplify assay preparation and analytical workflows.
  • High purity (98.0%) suitable for research applications.
  • Available as 10 mM solution in DMSO (1 mL) for convenient use.
  • Also available in solid quantities (100 mg-1 g) and bulk sizes on request.
  • Molecular weight 150.13 g/mol; formula C5H10O5.
  • Useful in studies of ATP metabolism, energy pathways, and protein glycation.