Learn More
Medchemexpress LLC HY-W018772 100mg Medchemexpress, D-Ribose(mixture of isomers) CAS:50-69-1 Purity:>98%
SDP
Catalog No. 501934646
Encompass_Preferred
Medchemexpress, HY-W018772 100mg D-Ribose(mixture of isomers) CAS:50-69-1 D-Ribose(mixture of isomers) is an energy enhancer, and acts as a sugar moiety of ATP, and widely used as a metabolic therapy supplement for chronic fatigue syndrome or cardiac energy metabolism. D-Ribose(mixture of isomers) is active in protein glycation, induces NF-κB inflammation in a RAGE-dependent manner [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4646 Supplier Medchemexpress LLC Supplier No. HYW018772100MG
Medchemexpress, HY-W018772 100mg D-Ribose(mixture of isomers) CAS:50-69-1 D-Ribose(mixture of isomers) is an energy enhancer, and acts as a sugar moiety of ATP, and widely used as a metabolic therapy supplement for chronic fatigue syndrome or cardiac energy metabolism. D-Ribose(mixture of isomers) is active in protein glycation, induces NF-κB inflammation in a RAGE-dependent manner [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.