Learn More
Sigma Aldrich Fine Chemicals Biosciences D Ribulose 15 Diphosphate
Catalog No. 502907654
Encompass
Exists as a 557AkDa hexadecamer composed of eight heavy chains each with a molecular weight of approximately 56AkDa and eight light chains of molecular weight 14AkDa. Each molecule contains one magnesium ion. pH optimum ~7.9. KM for CO2 ~0.45 mM. Ribulose diphosphate becomes inhibitory at concentrations exceeding 0.7 mM. Orthophosphate and ammonium sulfate are competitive inhibitors. 3-Phosphoglycerate is a noncompetitive inhibitor.
Catalog No. 50-290-7654 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R80005UN
Exists as a 557AkDa hexadecamer composed of eight heavy chains each with a molecular weight of approximately 56AkDa and eight light chains of molecular weight 14AkDa. Each molecule contains one magnesium ion. pH optimum ~7.9. KM for CO2 ~0.45 mM. Ribulose diphosphate becomes inhibitory at concentrations exceeding 0.7 mM. Orthophosphate and ammonium sulfate are competitive inhibitors. 3-Phosphoglycerate is a noncompetitive inhibitor.