Learn More
MSC Value Collection Polyurethane Coated Cut-Resistant Gloves, Size 7, ANSI A3, 1/Pr.
Catalog No. 5000685131
Encompass
These high-visibility safety gloves feature a form-fitting DuraBlend® shell designed to provide reliable hand protection in demanding work environments. Equipped with a polyurethane palm coating, these gloves offer enhanced grip and durability while maintaining comfort for extended wear. They are suitable for a variety of tasks including material handling and assembly.
  • ANSI cut level A3 for protection against sharp objects
  • ANSI abrasion resistance level 4
  • ANSI puncture resistance level 2
  • High-visibility color for increased awareness
  • Color-coded overcast facilitates easy sizing and reduces fraying
  • Machine washable design for easy cleaning
Catalog No. 50-006-85131 Supplier MSC Supplier No. 73709933
These high-visibility safety gloves feature a form-fitting DuraBlend® shell designed to provide reliable hand protection in demanding work environments. Equipped with a polyurethane palm coating, these gloves offer enhanced grip and durability while maintaining comfort for extended wear. They are suitable for a variety of tasks including material handling and assembly.
  • ANSI cut level A3 for protection against sharp objects
  • ANSI abrasion resistance level 4
  • ANSI puncture resistance level 2
  • High-visibility color for increased awareness
  • Color-coded overcast facilitates easy sizing and reduces fraying
  • Machine washable design for easy cleaning