Magid ROC GP500 NitriX Grip Palm Coated Gloves are designed for demanding work environments, featuring a grey 13-gauge polyester shell and a black NitriX Grip Technology palm coating. These gloves provide exceptional grip and protection, especially in oily conditions, while offering water-repellent properties.
- Grey 13-gauge 100% polyester shell
- Black NitriX Grip Technology palm (sandy nitrile coating)
- Exceptional grip and protection in oily conditions with water-repellent feature
- Seamless machine knit polyester shell for flexibility and comfort
- Great dexterity and tactile sensitivity
- Dark color coating hides dirt, extending glove life
- ANSI Cut Level 1 and ANSI Abrasion Level 4 resistance
- Knit wrist with colored overedge and rubber band finish
- Launderable for extended wear time