Learn More
Cole-Parmer Digi-Sense Stainless Steel Bimetal Pocket Thermometer, 1.75" Dial, Glass Lens, 8" Stem, -40 to 160F, 2F Div
Catalog No. 501961474 Shop All Cole-Parmer Products
Encompass_Preferred
Choose the dial size and temperature range on this shock resistant thermometer to meet your specific needs. Factory tested to three points with a 1% accuracy. Hermetically sealed dial stops internal fogging. Minimum immersion: liquids 2" (50 mm), gases 4" (100 mm). Comes with a 304 Stainless Steel dial case and stem.
Catalog No. 50-196-1474 Supplier Cole-Parmer Supplier No. 0808088
Choose the dial size and temperature range on this shock resistant thermometer to meet your specific needs. Factory tested to three points with a 1% accuracy. Hermetically sealed dial stops internal fogging. Minimum immersion: liquids 2" (50 mm), gases 4" (100 mm). Comes with a 304 Stainless Steel dial case and stem.