Learn More
Cole-Parmer Digi-Sense Stainless Steel Bimetal Pocket Thermometer, 2" Dial, Poly Lens, 8" Stem, 0 to 220F, 2F Div
Catalog No. 501961477 Shop All Cole-Parmer Products
Encompass_Preferred
Choose the dial size and temperature range on this shock resistant thermometer to meet your specific needs. Factory tested to three points with a 1% accuracy. Hermetically sealed dial stops internal fogging. Minimum immersion: liquids 2" (50 mm), gases 4" (100 mm). Comes with a 304 Stainless Steel dial case and stem.
Catalog No. 50-196-1477 Supplier Cole-Parmer Supplier No. 0808098
Choose the dial size and temperature range on this shock resistant thermometer to meet your specific needs. Factory tested to three points with a 1% accuracy. Hermetically sealed dial stops internal fogging. Minimum immersion: liquids 2" (50 mm), gases 4" (100 mm). Comes with a 304 Stainless Steel dial case and stem.