Learn More
Cole-Parmer Digi-Sense PFA Safety Coated Liquid-In-Glass Thermometer; 0 to 230F, 76mm Immersion, Organic Liquid Fill
Catalog No. 501961468 Shop All Cole-Parmer Products
Encompass_Preferred
These thermometers are ideal for use in laboratories, universities, food/beverage, environmental, wastewater, and many other general-purpose applications. In the event of breakage, the PTFE coating contains the broken liquid and glass, preventing contamination or injury. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered.
Catalog No. 50-196-1468 Supplier Cole-Parmer Supplier No. 0807777
These thermometers are ideal for use in laboratories, universities, food/beverage, environmental, wastewater, and many other general-purpose applications. In the event of breakage, the PTFE coating contains the broken liquid and glass, preventing contamination or injury. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered.