Learn More
Cole-Parmer Digi-Sense General Purpose Liquid-In-Glass Thermometer; -10 to 260C, 76mm Immersion, Organic Liquid Fill
Catalog No. 501961576 Shop All Cole-Parmer Products
Encompass_Preferred
These thermometers are inexpensive and safe for use in laboratories, universities, food/beverage, environmental, wastewater, petrochemical, pharmaceutical and many other general-purpose applications. Choose from partial immersion or total immersion thermometers. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include a ring top and a statement of accuracy supplied by manufacturer.
Catalog No. 50-196-1576 Supplier Cole-Parmer Supplier No. 9026005
These thermometers are inexpensive and safe for use in laboratories, universities, food/beverage, environmental, wastewater, petrochemical, pharmaceutical and many other general-purpose applications. Choose from partial immersion or total immersion thermometers. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include a ring top and a statement of accuracy supplied by manufacturer.