Learn More
Cole-Parmer Digi-Sense Armored Liquid-In-Glass Thermometer; 20 to 500F, 76mm Immersion, Organic Liquid Fill
Catalog No. 501961471 Shop All Cole-Parmer Products
Encompass_Preferred
Ideal for use in application where thermometers needs added protection from minor shocks. Nickel/plated brass armor shell protects these thermometers from accidental breakage during use and storage. The armor case has a screw cap with ring top and viewing window; the lower section that surrounds the bulb has small openings for circulation. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include statement of accuracy and a triangular-style antiroll fitting.
Catalog No. 50-196-1471 Supplier Cole-Parmer Supplier No. 0807798
Ideal for use in application where thermometers needs added protection from minor shocks. Nickel/plated brass armor shell protects these thermometers from accidental breakage during use and storage. The armor case has a screw cap with ring top and viewing window; the lower section that surrounds the bulb has small openings for circulation. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include statement of accuracy and a triangular-style antiroll fitting.