Learn More
Cole-Parmer Digi-Sense ASTM Like Liquid-In-Glass Thermometer; 9C / Low-Pensky-Martens, 57mm Immersion, -5 to 110C, Organic Liquid Fill
Catalog No. 501961458 Shop All Cole-Parmer Products
Encompass_Preferred
These thermometers are precision manufactured to conform to ASTM E-1 specifications except for not being mercury filled. Designed for use in quality control laboratories or when mercury filled thermometers can not be used but where ASTM standards are still needed. ASTM established thermometer standards suited for special applications, and assigned an ASTM number according to each specification. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered.
Catalog No. 50-196-1458 Supplier Cole-Parmer Supplier No. 0800907
These thermometers are precision manufactured to conform to ASTM E-1 specifications except for not being mercury filled. Designed for use in quality control laboratories or when mercury filled thermometers can not be used but where ASTM standards are still needed. ASTM established thermometer standards suited for special applications, and assigned an ASTM number according to each specification. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered.