Learn More
Cole-Parmer Digi-Sense ASTM S63F-03 Individually Calibrated Liquid-In-Glass Thermometer; 18/89F, Organic Liquid Fill
Catalog No. 501961461 Shop All Cole-Parmer Products
Encompass_Preferred
These NIST-traceable thermometers fulfill requirements for performing ASTM methods without the worry of mercury contamination. Organic fill solution is safer to clean up in the event of breakage. The calibration report states corrections to one-tenth of the smallest division, providing an accurate offset for ensuring precise measurement records.
Catalog No. 50-196-1461 Supplier Cole-Parmer Supplier No. 0800967
These NIST-traceable thermometers fulfill requirements for performing ASTM methods without the worry of mercury contamination. Organic fill solution is safer to clean up in the event of breakage. The calibration report states corrections to one-tenth of the smallest division, providing an accurate offset for ensuring precise measurement records.