Learn More
Cole-Parmer Digi-Sense Incubator Verification Thermometer; 25 to 45C
Supplier:  Cole-Parmer 0807734
Encompass_Preferred
Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. All thermometers are supplied in a glass bead filled unbreakable plastic bottle and can be removed from the bottle for recalibration. Use the magnetic attachment with pressure-sensitive backing to attach the bottle to a metal wall or door. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an statement of accuracy and a triangular-style antiroll fitting.
Catalog No. 50-196-1467