Learn More
Santa Cruz Biotechnology D-SACCHARIC ACID 1,4 LACTONE
Supplier:  Santa Cruz Biotechnology SC221521A
Encompass
D-Saccharic acid 1,4-lactone monohydrate inhibits beta-glucuronidase from Littorina littorea. 250mg. 61278-30-6.
Catalog No. NC0858864