Learn More
Sigma Aldrich Fine Chemicals Biosciences D Saccharic acid potassium100G
Catalog No. 502908069
Encompass
D-Saccharic acid potassium salt is present in plants and animals. It is produced by the chemical oxidation of glucose with nitric acid. D-Saccharic acid lowers cholesterol and has chemotherapeutic property. It also has antimutagenic activity.
Catalog No. 50-290-8069 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S4140100G
Add to Cart
Add to Cart
D-Saccharic acid potassium salt is present in plants and animals. It is produced by the chemical oxidation of glucose with nitric acid. D-Saccharic acid lowers cholesterol and has chemotherapeutic property. It also has antimutagenic activity.