Learn More
Lonza Walkersville D-SAEC-COPD - COPD
Catalog No. 5000496435
Encompass
Chronic bronchitis is marked by long-term cough and excess mucous Emphysema leads to the destruction of the lungs over time Most people with COPD have a combination of both conditions Usually COPD is associated with bronchial epithelium changes Lonza s Human Small Airway Epithelial Cells (SAEC) with COPD are isolated from diseased human lung tissue from COPD donors Cells are isolated from the distal portion of the lung in the 1mm bronchiole area All cells test negative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B andhepatitis C are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased
Catalog No. 50-004-96435 Supplier Lonza Walkersville Supplier No. CC2934
Chronic bronchitis is marked by long-term cough and excess mucous Emphysema leads to the destruction of the lungs over time Most people with COPD have a combination of both conditions Usually COPD is associated with bronchial epithelium changes Lonza s Human Small Airway Epithelial Cells (SAEC) with COPD are isolated from diseased human lung tissue from COPD donors Cells are isolated from the distal portion of the lung in the 1mm bronchiole area All cells test negative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B andhepatitis C are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased