Learn More
AdipoGen D-(-)-Salicin (100 g)
SDP
Supplier:  AdipoGen CDXS0001G100
Encompass_Preferred
Chemical. CAS: 138-52-3. Formula: C13H18O7. MW: 286.28. Salicin is an alcoholic beta-glucoside derived from willow bark that produces antioxidant and anti-inflammatory (including analgesic and antipyretic) effects very similar to that of aspirin. It acts as a non-selective COX inhibitor for COX-1 and COX-2. It has been used in the treatment of rheumatism. Shown to inhibit angiogenesis by blocking the ROS-ERK pathways and to have in vivo anticancer activity.
Catalog No. 50-205-4635