Learn More
Apexbio Technology LLC D-(-)-Salicin 138-52-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094423
Encompass_Preferred
D-(-)-Salicin (CAS 138-52-3) is a -glucoside compound isolated from the purple willow (Salix spp ) Upon enzymatic hydrolysis it releases saligenin which is subsequently oxidized to salicylic acid a known modulator of inflammatory pathways through cyclooxygenase (COX) inhibition D-(-)-Salicin is utilized in biomedical research to investigate anti-inflammatory mechanisms pain modulation and metabolic conversion of glycosides It is soluble in boiling water alkaline solutions pyridine and acetic acid For optimal stability the compound should be stored in a sealed container under cool dry conditions
Catalog No. 50-000-94423 Supplier Apexbio Technology LLC Supplier No. N18805.1
D-(-)-Salicin (CAS 138-52-3) is a -glucoside compound isolated from the purple willow (Salix spp ) Upon enzymatic hydrolysis it releases saligenin which is subsequently oxidized to salicylic acid a known modulator of inflammatory pathways through cyclooxygenase (COX) inhibition D-(-)-Salicin is utilized in biomedical research to investigate anti-inflammatory mechanisms pain modulation and metabolic conversion of glycosides It is soluble in boiling water alkaline solutions pyridine and acetic acid For optimal stability the compound should be stored in a sealed container under cool dry conditions