Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp D Salicin 500 G
SDP

Catalog No. 502114044
Encompass_Preferred

CAS Number: 138-52-3
Molecular Formula: C13H18O7
Molecular Weight: 286.3
Chemical Purity: >99%
Soluble in: Water
Store at: Room Temperature

Catalog No. 50-211-4044 Supplier Research Products International Corp Supplier No. S23200500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS Number: 138-52-3
Molecular Formula: C13H18O7
Molecular Weight: 286.3
Chemical Purity: >99%
Soluble in: Water
Store at: Room Temperature

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.