Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(-)-Salicin, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013027 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3027 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3027 Supplier MilliporeSigma Supelco Supplier No. 79588100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical standard for use in chromatography applications.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Salicin
CAS 138-52-3
Physical Form Neat
Linear Formula C13H18O7
Technique Gas Chromatography
MDL Number MFCD00006590
Synonym 2-(Hydroxymethyl)phenyl-beta-D-glucopyranoside; Salicoside; Salicyl alcohol glucoside; Saligenin beta-D-glucoside
Type Analytical Standard
Molecular Weight (g/mol) 286.28
Quantity 100 mg
Formula Weight 286.28
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.