Learn More
Medchemexpress LLC D-Sedoheptulose-7-phosphate barium | 17187-72-3 | 99.70% | 427.49 | 1 MG
SDP
Catalog No. 5000436885
Encompass_Preferred
D-Sedoheptulose-7-phosphate barium is a common precursor for the heptoses of septacidin (group III) and hygromycin B (group IV). It can be converted to NDP-heptoses through similar biosynthetic pathways in those compounds.
  • Common precursor for heptoses of septacidin (group III) and hygromycin B (group IV)
  • Converts to NDP-heptoses through similar biosynthetic pathways
Catalog No. 50-004-36885 Supplier Medchemexpress LLC Supplier No. HY113206A1MG
D-Sedoheptulose-7-phosphate barium is a common precursor for the heptoses of septacidin (group III) and hygromycin B (group IV). It can be converted to NDP-heptoses through similar biosynthetic pathways in those compounds.
  • Common precursor for heptoses of septacidin (group III) and hygromycin B (group IV)
  • Converts to NDP-heptoses through similar biosynthetic pathways