Learn More
AdipoGen D-Selenocystine
SDP
Supplier:  AdipoGen CDXS0117M025
Encompass_Preferred
Chemical. CAS 26932-45-6. Formula C6H12N2O4Se2. MW 334.09. Synthetic. D-isoform of the 21st proteinogenic amino acid L-selenocystine, which is a natural thioredoxin reductase inhibitor. Shows minimal ability to induce GPx activity.
Catalog No. 50-256-0061