Learn More
Medchemexpress LLC (D-Ser1)-ACTH (1-24) (human, bovine, mouse, ovine, porcine, rabbit, rat) | 18067-65-7 | 2933.44 | 1 MG
SDP
Catalog No. 5000397399
Encompass_Preferred
This product is an adrenocorticotropic hormone (ACTH) analogue, identified as (D-Ser1)-ACTH (1-24) for human, bovine, mouse, ovine, porcine, rabbit, and rat species. It targets the Melanocortin Receptor and is involved in GPCR/G Protein and Neuronal Signaling pathways. This product is for research use only and has not been fully validated for medical applications.
  • Adrenocorticotropic hormone (ACTH) analogue
  • Targets melanocortin receptor
  • Involved in GPCR/G protein pathway
  • Involved in neuronal signaling
  • Suitable for research applications
Catalog No. 50-003-97399 Supplier Medchemexpress LLC Supplier No. HYP35641MG
This product is an adrenocorticotropic hormone (ACTH) analogue, identified as (D-Ser1)-ACTH (1-24) for human, bovine, mouse, ovine, porcine, rabbit, and rat species. It targets the Melanocortin Receptor and is involved in GPCR/G Protein and Neuronal Signaling pathways. This product is for research use only and has not been fully validated for medical applications.
  • Adrenocorticotropic hormone (ACTH) analogue
  • Targets melanocortin receptor
  • Involved in GPCR/G protein pathway
  • Involved in neuronal signaling
  • Suitable for research applications