Learn More
Medchemexpress LLC (D-Ser1)-ACTH (1-24) (human, bovine, mouse, ovine, porcine, rabbit, rat) | 18067-65-7 | >99.9% | 2933.44 | 5 MG
SDP
Catalog No. 5000396723
Encompass_Preferred
(D-Ser1)-ACTH (1-24) is an adrenocorticotropic hormone (ACTH) analogue. It requires specific storage conditions to maintain stability.
  • Suitable for sealed storage, away from moisture.
  • Powder can be stored at -80°C for 2 years or -20°C for 1 year.
  • In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-003-96723 Supplier Medchemexpress LLC Supplier No. HYP35645MG
(D-Ser1)-ACTH (1-24) is an adrenocorticotropic hormone (ACTH) analogue. It requires specific storage conditions to maintain stability.
  • Suitable for sealed storage, away from moisture.
  • Powder can be stored at -80°C for 2 years or -20°C for 1 year.
  • In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month.