Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-serine-2,3,3-d3 | 1414348-52-9 | 96.4% | 108.11 g/mol | C3H4D3NO3 | 1 MG
SDP

Catalog No. 5000212399
Encompass_Preferred

D-Serine-d3 is a deuterium-labeled form of the endogenous amino acid D-serine, provided as an isotopic reference standard for analytical and biochemical research. It is commonly used in mass spectrometry, tracer studies, and assays probing NMDA receptor-related neurotransmission and synaptic processes.

  • Deuterium-labeled (d3) analogue suitable for isotope-dilution analyses.
  • High purity (96.4%) for analytical reproducibility.
  • Molecular weight 108.11 g/mol and formula C3H4D3NO3.
  • Supplied in 1 mg quantity for standard preparation.
  • Supports studies of NMDA receptor-mediated neurotransmission and synaptic plasticity.

Catalog No. 50-002-12399 Supplier Medchemexpress LLC Supplier No. HY100808S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Serine-d3 is a deuterium-labeled form of the endogenous amino acid D-serine, provided as an isotopic reference standard for analytical and biochemical research. It is commonly used in mass spectrometry, tracer studies, and assays probing NMDA receptor-related neurotransmission and synaptic processes.

  • Deuterium-labeled (d3) analogue suitable for isotope-dilution analyses.
  • High purity (96.4%) for analytical reproducibility.
  • Molecular weight 108.11 g/mol and formula C3H4D3NO3.
  • Supplied in 1 mg quantity for standard preparation.
  • Supports studies of NMDA receptor-mediated neurotransmission and synaptic plasticity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.