Learn More
Medchemexpress LLC D-serine | 312-84-5 | MFCD00004269 | >=98.0% | 105.09 g/mol | C3H7NO3 | 5 G
SDP
Catalog No. 5000211182
Encompass_Preferred
D-Serine is the R-enantiomer of serine and an endogenous amino acid that functions as a co-agonist at N-methyl-D-aspartate (NMDA) receptors. It is supplied as a research-grade solid for use in neuroscience and biochemical studies of NMDAR-mediated neurotransmission, synaptic plasticity, and neurotoxicity.
  • Endogenous NMDA receptor co-agonist.
  • Suitable for neuroscience and biochemical research.
  • High purity (≥98%).
  • Molecular weight 105.09 g/mol.
  • Supplied as a solid powder for analytical and research applications.
Catalog No. 50-002-11182 Supplier Medchemexpress LLC Supplier No. HY1008085G
D-Serine is the R-enantiomer of serine and an endogenous amino acid that functions as a co-agonist at N-methyl-D-aspartate (NMDA) receptors. It is supplied as a research-grade solid for use in neuroscience and biochemical studies of NMDAR-mediated neurotransmission, synaptic plasticity, and neurotoxicity.
  • Endogenous NMDA receptor co-agonist.
  • Suitable for neuroscience and biochemical research.
  • High purity (≥98%).
  • Molecular weight 105.09 g/mol.
  • Supplied as a solid powder for analytical and research applications.