Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-serine, N-[[2-[(3-cyanophenyl)methoxy]-4-...]] hydrochloride (1:1) | 2113650-04-5 | 98.0% | 50 MG
SDP

Catalog No. 5000231185
Encompass_Preferred

BMS-1001 hydrochloride is a small-molecule inhibitor of the PD-1/PD-L1 interaction supplied as the hydrochloride salt for research use. It is described as orally active and exhibits low cellular toxicity, and is provided as a solid with high purity suitable for cellular assays and interaction studies.

  • Orally active PD-1/PD-L1 interaction inhibitor.
  • High purity (98.0%).
  • Molecular weight 631.11 g·mol⁻1.
  • Solid appearance; store sealed and away from moisture.
  • Storage in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Available in small milligram pack sizes for research use.

Catalog No. 50-002-31185 Supplier Medchemexpress LLC Supplier No. HY12063550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-1001 hydrochloride is a small-molecule inhibitor of the PD-1/PD-L1 interaction supplied as the hydrochloride salt for research use. It is described as orally active and exhibits low cellular toxicity, and is provided as a solid with high purity suitable for cellular assays and interaction studies.

  • Orally active PD-1/PD-L1 interaction inhibitor.
  • High purity (98.0%).
  • Molecular weight 631.11 g·mol⁻1.
  • Solid appearance; store sealed and away from moisture.
  • Storage in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Available in small milligram pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.