Learn More
Medchemexpress LLC D-serine, N-[[2-[(3-cyanophenyl)methoxy]-4-...]] hydrochloride | 2113650-04-5 | 98.0% | 100 MG
SDP
Catalog No. 5000232072
Encompass_Preferred
BMS-1001 hydrochloride is a small-molecule inhibitor of the PD-1/PD-L1 interaction supplied as the hydrochloride salt for research use. It shows potent binding activity and is intended for in vitro studies of immune checkpoint pathways.
  • Orally active PD-1/PD-L1 inhibitor with low cellular toxicity.
  • IC50 of 2.25 nM in HTRF binding assay.
  • Molecular weight 631.11 and formula C35H35ClN2O7.
  • High purity (98.0%) suitable for research applications.
  • Provided as a hydrochloride salt to aid solubility in assay buffers.
Catalog No. 50-002-32072 Supplier Medchemexpress LLC Supplier No. HY120635100MG
BMS-1001 hydrochloride is a small-molecule inhibitor of the PD-1/PD-L1 interaction supplied as the hydrochloride salt for research use. It shows potent binding activity and is intended for in vitro studies of immune checkpoint pathways.
  • Orally active PD-1/PD-L1 inhibitor with low cellular toxicity.
  • IC50 of 2.25 nM in HTRF binding assay.
  • Molecular weight 631.11 and formula C35H35ClN2O7.
  • High purity (98.0%) suitable for research applications.
  • Provided as a hydrochloride salt to aid solubility in assay buffers.