Learn More
Medchemexpress LLC D-Sorbitol-13C | 287100-73-6 | 98.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0400S11MG
Encompass_Preferred
D-Sorbitol-13C is the 13C labeled D-Sorbitol. D-Sorbitol (Sorbitol) is a six-carbon sugar alcohol that can be used as a sugar substitute. It can also be used as a stabilizing excipient and/or isotonicity agent, sweetener, humectant, thickener, and dietary supplement.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06433