Learn More
Medchemexpress LLC D-Sorbitol-13C | 287100-73-6 | 98.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0400S15MG
Encompass_Preferred
D-Sorbitol-13C is the 13C labeled D-Sorbitol, a six-carbon sugar alcohol used as a sugar substitute. It functions as a stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement.
  • Tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05427