Learn More
Medchemexpress LLC D-Sorbitol-13C6 | 121067-66-1 | 99.9% | 5 MG
SDP
Catalog No. 5000407132
Encompass_Preferred
D-Sorbitol-13C6 is the 13C labeled D-Sorbitol, a six-carbon sugar alcohol used as a sugar substitute. It functions as a stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement. This compound can be utilized as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS, leveraging stable heavy isotopes like carbon-13 incorporated into drug molecules for quantitation during drug development.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07132 Supplier Medchemexpress LLC Supplier No. HYB0400S165MG
D-Sorbitol-13C6 is the 13C labeled D-Sorbitol, a six-carbon sugar alcohol used as a sugar substitute. It functions as a stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement. This compound can be utilized as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS, leveraging stable heavy isotopes like carbon-13 incorporated into drug molecules for quantitation during drug development.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS